missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetoacetate, 99%, pure, Thermo Scientific Chemicals

Catalog Number 10449220
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic drum
141-97-9
C6H10O3
130.14
MFCD00009199
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
8868
CHEBI:4893
ethyl 3-oxobutanoate
CCOC(=O)CC(=O)C
141-97-9
C6H10O3
130.14
MFCD00009199
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
8868
CHEBI:4893
ethyl 3-oxobutanoate
CCOC(=O)CC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -45.0°C
Color Colorless
Density 1.0300g/mL
Boiling Point 180.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula CH3COCH2CO2C2H5
Refractive Index 1.4180 to 1.4200
Quantity 25 L
Beilstein 03, 632
Fieser 12,222
Merck Index 15, 3812
Specific Gravity 1.03
Solubility Information Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone
Viscosity 1.696 mPa.s (20°C)
Formula Weight 130.14
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethyl acetoacetate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'