missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate (Sequencing), Fisher BioReagents

Catalog Number p-25739
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
This item is not returnable. View return policy
141-78-6
C4H8O2
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
This solvent is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY
Melting Point -83.5°C
Absorbance 1 max. at 255nm, 0.15 max. at 260nm, 0.025 max. at 270nm
Color Colorless
Density 0.902g/cm³
Boiling Point 75°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 10ppm max.
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOCH2CH3
Vapor Pressure 103mbar at 20°C
UN Number 1173
Viscosity 0.45 cP at 20°C
Water 0.02% max.
Merck Index 15, 3811
Formula Weight 88.11
Percent Purity ≥99.9%
Grade Sequencing
Chemical Name or Material Ethyl Acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'