missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, for HPLC, Fisher Chemical™

Catalog Number 10040140
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
141-78-6
C4H8O2
88.106
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
CCOC(=O)C
141-78-6
C4H8O2
88.106
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
CCOC(=O)C

C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C

TRUSTED_SUSTAINABILITY
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Color Colorless
IUPAC Name ethyl acetate
Formula Weight 88.11g/mol
Vapor Pressure 103mbar at 20°C
Viscosity 0.45cp at 20°C
Vapor Density 3.04
Melting Point -83.5°C
Boiling Point 75°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'