missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Specifications
CAS | 141-78-6 |
Molecular Formula | C4H8O2 |
Synonym | ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
SMILES | CCOC(=O)C |
IUPAC Name | ethyl acetate |
Molecular Weight (g/mol) | 88.106 |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
Formula Weight | 88.11 |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'