missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10386370
48.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


48.98 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
141-78-6
C4H8O2
88.106
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C
141-78-6
C4H8O2
88.106
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
8857
CHEBI:27750
ethyl acetate
CCOC(=O)C

Ethyl Acetate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point -83.5°C
Color Colorless
Boiling Point 75°C
Quantity 5 L
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Viscosity 0.45cp at 20°C
Formula Weight 88.11g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'