missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10182392
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
141-78-6
C4H8O2
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
ethyl acetate
141-78-6
C4H8O2
9171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
CHEBI:27750
ethyl acetate

C4H8O2, CAS Number-141-78-6, acetic acid ethyl ester, acetic acid, ethyl ester, acetic ester, acetic ether, acetidin, acetoxyethane, ethylacetate, ethyl acetic ester, ethyl ethanoate, vinegar naphtha, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate

TRUSTED_SUSTAINABILITY
Melting Point -83.5°C
Color Colorless
Boiling Point 75°C
Quantity 5 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Viscosity 0.45cp at 20°C
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
SMILES CCOC(=O)C
Molecular Weight (g/mol) 88.106
Formula Weight 88.11g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P240-Ground/bond container and receiving equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'