Learn More
ethyl 7-chloropyrazolo[1,5-a]pyrimidine-5-carboxylate, Maybridge
Brand: Maybridge MO08583DA
Code : 31
Additional Details : CAS Number : 61098-37-1 Weight : 0.00100kg
Quantity | 1g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
ethyl 7-chloropyrazolo[1, 5-a]pyrimidine-5-carboxylate | |
97% | |
C9H8ClN3O2 | |
225.63 |
Specifications
ethyl 7-chloropyrazolo[1, 5-a]pyrimidine-5-carboxylate | |
Amber Glass Bottle | |
C9H8ClN3O2 | |
225.63 |
61098-37-1 | |
97% | |
1g |