Learn More
Thermo Scientific Maybridge Ethyl 7-chloropyrazolo[1,5-a]pyrimidine-5-carboxylate, 5g, 97%
Brand: Thermo Scientific Maybridge MO08583DE
Ethyl 7-chloropyrazolo[1,5-a]pyrimidine-5-carboxylate, 5g, 97%
Code : 31
Additional Details : CAS Number : 61098-37-1 Weight : 0.00500kg