missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloropyridine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7053928
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21190-89-6
C8H8ClNO2
185.607
MFCD09878438
ORSVWYSFUABOQU-UHFFFAOYSA-N
ethyl 6-chloropicolinate, ethyl 6-chloro-2-pyridinecarboxylate, 6-chloropyridine-2-carboxylic acid ethyl ester, 6-chloro-pyridine-2-carboxylic acid ethyl ester, 2-pyridinecarboxylicacid, 6-chloro-, ethyl ester, 2-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem17513, acmc-209fib, 2-chloro-6-ethoxycarbonyl-pyridine
11063114
ethyl 6-chloropyridine-2-carboxylate
CCOC(=O)C1=NC(=CC=C1)Cl
This item is not returnable. View return policy
21190-89-6
C8H8ClNO2
185.607
MFCD09878438
ORSVWYSFUABOQU-UHFFFAOYSA-N
ethyl 6-chloropicolinate, ethyl 6-chloro-2-pyridinecarboxylate, 6-chloropyridine-2-carboxylic acid ethyl ester, 6-chloro-pyridine-2-carboxylic acid ethyl ester, 2-pyridinecarboxylicacid, 6-chloro-, ethyl ester, 2-pyridinecarboxylic acid, 6-chloro-, ethyl ester, pubchem17513, acmc-209fib, 2-chloro-6-ethoxycarbonyl-pyridine
11063114
ethyl 6-chloropyridine-2-carboxylate
CCOC(=O)C1=NC(=CC=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Formula Weight 185.61
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Ethyl 6-chloropyridine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'