Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3h-dibenzo[f,ij]Isoquinolinecarboxylate, 98%, ACROS Organics™


Overview
£8.68 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 374020010

1GR Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3H-dibenzo¢f,ij! isoquinolinecarboxylate, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 51418-86-1 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10422303

Quantity Price
1 £ 21.7 / 1g
EU Stock 2
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3H-dibenzo[f,ij] isoquinolinecarboxylate
Assay Percent Range 98%
Molecular Formula C20H14BrNO4
Formula Weight 412.23
MDL Number MFCD02728506

Specification

Physical Form Crystalline Powder
Formula Weight 412.23
Quantity 1g
Color Brown
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 51418-86-1
Assay Percent Range 98%
MDL Number MFCD02728506
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Elemental Analysis 97.5% min.
Molecular Formula C20H14BrNO4
Chemical Name or Material Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3H-dibenzo[f,ij] isoquinolinecarboxylate
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Special Offers