missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 5-nitro-1-indoleacetate, 97%

Brand:  Alfa Aesar™ H63509.03

Code : NEW

Additional Details : CAS Number : 226901-49-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400249

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 5-nitro-1-indoleacetate
248.23
11123712
226901-49-1
MFCD11543812
5-Nitro-1-indoleacetic acid ethyl ester

Specifications

Ethyl 5-nitro-1-indoleacetate
226901-49-1
97%
MFCD11543812
5-Nitro-1-indoleacetic acid ethyl ester
1g
Solid
248.23
11123712
Videos
SDS