missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate, 95%

Brand:  Alfa Aesar™ H63751.03

Code : NEW

Additional Details : CAS Number : 2199-59-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458496

  • £117.20 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate
195.22
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid ethyl ester
2199-59-9
MFCD00030352

Specifications

Ethyl 5-formyl-2,4-dimethylpyrrole-3-carboxylate
Solid
195.22
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid ethyl ester
2199-59-9
95%
MFCD00030352
1g
Videos
SDS