missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog Number 15427828
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1197193-30-8
C7H7BrN2O2
231.049
MFCD12964050
CZQFHUXIWZYQGP-UHFFFAOYSA-N
ethyl 5-bromopyrimidine-2-carboxyate, ethyl-5-bromopyrimidine-2-carboxylate, 5-bromo-2-pyrimidinecarboxylic acid ethyl ester, ethyl5-bromopyrimidine-2-carboxylate, 5-bromopyrimidine-2-carboxylic acid ethyl ester
52987579
ethyl 5-bromopyrimidine-2-carboxylate
CCOC(=O)C1=NC=C(C=N1)Br
This item is not returnable. View return policy
1197193-30-8
C7H7BrN2O2
231.049
MFCD12964050
CZQFHUXIWZYQGP-UHFFFAOYSA-N
ethyl 5-bromopyrimidine-2-carboxyate, ethyl-5-bromopyrimidine-2-carboxylate, 5-bromo-2-pyrimidinecarboxylic acid ethyl ester, ethyl5-bromopyrimidine-2-carboxylate, 5-bromopyrimidine-2-carboxylic acid ethyl ester
52987579
ethyl 5-bromopyrimidine-2-carboxylate
CCOC(=O)C1=NC=C(C=N1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 231.05
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Ethyl 5-bromopyrimidine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'