missing translation for 'onlineSavingsMsg'
Learn More

Ethyl(5-bromobenzofuran)-2-carboxylate, 95%, Maybridge

Brand:  Maybridge CC31223DE

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 84102-69-2 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10153914

  • £174.00 / 5g


Estimated Shipment date
from Supplier 26-09-2017
Add to basket

Specifications

Chemical Identifiers

Ethyl (5-bromobenzofuran)-2-carboxylate
95%
269.09
95%
C11H9BrO3

Specifications

Ethyl (5-bromobenzofuran)-2-carboxylate
Amber Glass Bottle
95%
5g
84102-69-2
95%
C11H9BrO3
269.09
Documentation

Technical Documents