missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-1-methylpyrazole-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-4509963
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
31037-02-2
C7H11N3O2
169.18
MFCD00051652
MEUSJJFWVKBUFP-UHFFFAOYSA-N
ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
271299
CCOC(=O)C1=C(N)N(C)N=C1
This item is not returnable. View return policy
31037-02-2
C7H11N3O2
169.18
MFCD00051652
MEUSJJFWVKBUFP-UHFFFAOYSA-N
ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
271299
CCOC(=O)C1=C(N)N(C)N=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'