missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 277.15 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'