missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5,5-dimethyl-2,4-dioxohexanoate, 97%, Thermo Scientific™

Catalog Number p-4510606
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
13395-36-3
C10H16O4
200.234
NIMKIMUBJFWPTD-UHFFFAOYSA-N
ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
83403
ethyl 5,5-dimethyl-2,4-dioxohexanoate
CCOC(=O)C(=O)CC(=O)C(C)(C)C
This item is not returnable. View return policy
13395-36-3
C10H16O4
200.234
NIMKIMUBJFWPTD-UHFFFAOYSA-N
ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
83403
ethyl 5,5-dimethyl-2,4-dioxohexanoate
CCOC(=O)C(=O)CC(=O)C(C)(C)C
Assay Percent Range 97%
Quantity 1 g
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Ethyl 5, 5-dimethyl-2, 4-dioxohexanoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'