missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 4-(trifluoromethyl)phenylacetate, 95%

Brand:  Alfa Aesar™ H64099.03

Code : NEW

Additional Details : CAS Number : 721-63-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461139

  • £73.20 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 4-(trifluoromethyl)phenylacetate
232.2
4-(Trifluoromethyl)phenylacetic acid ethyl ester
721-63-1
MFCD07779351

Specifications

Ethyl 4-(trifluoromethyl)phenylacetate
1g
Solid
232.2
4-(Trifluoromethyl)phenylacetic acid ethyl ester
Colorless
721-63-1
95%
MFCD07779351
Videos
SDS