missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 4-methylthiazole-5-carboxylate, 95%

Brand:  Alfa Aesar™ H61142.03

Code : NEW

Additional Details : CAS Number : 20582-55-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437648

  • £172.14 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 4-methylthiazole-5-carboxylate
171.22
4-Methylthiazole-5-carboxylic acid ethyl ester
20582-55-2
MFCD00626873

Specifications

Ethyl 4-methylthiazole-5-carboxylate
1g
Liquid
171.22
4-Methylthiazole-5-carboxylic acid ethyl ester
Pale yellow
20582-55-2
95%
MFCD00626873
Videos
SDS