missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-iodobenzoate, 97%, Thermo Scientific Chemicals

Catalog Number p-130205
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
51934-41-9
C9H9IO2
276.07
MFCD00017344
YCBJOQUNPLTBGG-UHFFFAOYSA-N
p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
142891
ethyl 4-iodobenzoate
CCOC(=O)C1=CC=C(I)C=C1
This item is not returnable. View return policy
51934-41-9
C9H9IO2
276.07
MFCD00017344
YCBJOQUNPLTBGG-UHFFFAOYSA-N
p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
142891
ethyl 4-iodobenzoate
CCOC(=O)C1=CC=C(I)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.6500g/mL
Boiling Point 281.0°C to 283.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4CO2CH2CH3
Refractive Index 1.5865 to 1.5885
Quantity 5 mL
Specific Gravity 1.65
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in non-polar solvents,soluble in polar solvents
Formula Weight 276.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4-iodobenzoate, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'