missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-fluorobenzoate, 99%, Thermo Scientific Chemicals

Catalog Number 11457024
11.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
451-46-7
C9H9FO2
168.167
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester, ethyl p-fluorobenzoate, benzoic acid, 4-fluoro-, ethyl ester, p-fluorobenzoic acid, ethyl ester, ethyl4-fluorobenzoate, 4-fluoro-benzoic acid ethyl ester, benzoic acid, p-fluoro-, ethyl ester, ethyep-fluorobenzoate, pubchem3471, ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 25°C to 27°C
Density 1.146
Boiling Point 209°C to 210°C
Flash Point 81°C (177°F)
Odor Recognizable
Refractive Index 1.487
Quantity 5 g
Beilstein 1866794
Merck Index 14,4171
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'