missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chlorophenylacetate, 98+%, Thermo Scientific Chemicals

Catalog Number p-3523443
13.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14062-24-9
MFCD00018791
UTWBWFXECVFDPZ-UHFFFAOYSA-N
ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p
84177
ethyl 2-(4-chlorophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14062-24-9
MFCD00018791
UTWBWFXECVFDPZ-UHFFFAOYSA-N
ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p
84177
ethyl 2-(4-chlorophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Low Melting Solid
Assay Percent Range 98+%
Molecular Formula C10H11ClO2
Linear Formula ClC6H4CH2C(O)OCH2CH3
Molecular Weight (g/mol) 198.646
Formula Weight 198.65
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-chlorophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-chlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'