missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromophenylacetate, 99%, Thermo Scientific Chemicals

Catalog Number p-3593427
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14062-25-0
C10H11BrO2
243.1
MFCD00016333
ZFDCWHPNBWPPHG-UHFFFAOYSA-N
ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw
7020609
ethyl 2-(4-bromophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)Br
This item is not returnable. View return policy
14062-25-0
C10H11BrO2
243.1
MFCD00016333
ZFDCWHPNBWPPHG-UHFFFAOYSA-N
ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw
7020609
ethyl 2-(4-bromophenyl)acetate
CCOC(=O)CC1=CC=C(C=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29°C to 31°C
Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2OC(O)CH2C6H4Br
Quantity 1 g
Formula Weight 243.1
Percent Purity 99%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Ethyl 4-bromophenylacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-bromophenylacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'