Learn More
Ethyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.
Solubility
Soluble in ethanol, chloroform, ethyl ether and dilute acids. Sparingly soluble in water
Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases.
Specifications
Melting Point | 88°C to 92°C |
Boiling Point | 310°C |
Flash Point | >110°C (230°F) |
Odor | Odorless |
Quantity | 250 g |
Beilstein | 638434 |
Merck Index | 14,1086 |
Solubility Information | Soluble in ethanol,chloroform,ethyl ether and dilute acids. Sparingly soluble in water |
Formula Weight | 165.19 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only