missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4,6-dichloronicotinate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
5 g
Description
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.
Solubility
Soluble in methanol.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Ethyl 4,6-dichloronicotinate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a fine chemical intermediate.
Solubility
Soluble in methanol.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Color | Yellow |
Quantity | 250 mg |
Solubility Information | Soluble in methanol. |
Formula Weight | 220.05 |
Percent Purity | 95% |
Physical Form | Liquid |
Chemical Name or Material | Ethyl 4,6-dichloronicotinate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'