missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-2691436
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
372-31-6
C6H7F3O3
184.11
MFCD00000424
OCJKUQIPRNZDTK-UHFFFAOYSA-N
ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
67793
ethyl 4,4,4-trifluoro-3-oxobutanoate
CCOC(=O)CC(=O)C(F)(F)F
This item is not returnable. View return policy
372-31-6
C6H7F3O3
184.11
MFCD00000424
OCJKUQIPRNZDTK-UHFFFAOYSA-N
ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
67793
ethyl 4,4,4-trifluoro-3-oxobutanoate
CCOC(=O)CC(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -39.0°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 131.0°C (709.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CF3COCH2CO2C2H5
Refractive Index 1.3750 to 1.3780
Quantity 25 mL
Beilstein 03, II, 425
Specific Gravity 1.25
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: miscible with ethanol and ether
Formula Weight 184.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4, 4, 4-trifluoroacetoacetate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4,4,4-trifluoroacetoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'