missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate, 97%

Brand:  Alfa Aesar™ H33599.MD

Code : NEW

Additional Details : CAS Number : 129768-30-5 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15428545

  • £31.40 / 250mg


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Description

Description

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate
208.14
3-Trifluoromethyl-1H-pyrazole-5-carboxylic acid ethyl ester
129768-30-5
MFCD04111238

Specifications

Ethyl 3-trifluoromethyl-1H-pyrazole-5-carboxylate
93° to 98°C
129768-30-5
97%
MFCD04111238
White
250mg
Solid
208.14
3-Trifluoromethyl-1H-pyrazole-5-carboxylic acid ethyl ester
Videos
SDS