missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-(tert-butyl)-1-methyl-1H-pyrazole-5-carboxylate, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Quantity | 1 g |
Synonym | ethyl 3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylate, ethyl 3-tert-butyl-1-methylpyrazole-5-carboxylate, 1h-pyrazole-5-carboxylicacid, 3-1,1-dimethylethyl-1-methyl-, ethyl ester, acmc-1c1ez, ethyl3-tert-butyl-1-methylpyrazole-5-carboxylate, 5-tert-butyl-2-methyl-pyrazole-3-carboxylic acid ethyl ester, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid ethyl ester |
Formula Weight | 210 |
Chemical Name or Material | Ethyl 3-(tert-butyl)-1-methyl-1H-pyrazole-5-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'