missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number p-131675
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
121-39-1
C11H12O3
192.21
MFCD00005123
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
8469
ethyl 3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)C2=CC=CC=C2
This item is not returnable. View return policy
121-39-1
C11H12O3
192.21
MFCD00005123
GOMAKLPNAAZVCJ-UHFFFAOYSA-N
8469
ethyl 3-phenyloxirane-2-carboxylate
CCOC(=O)C1C(O1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 96.0°C (0.5 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5165 to 1.5205
Quantity 5 mL
Beilstein 18,42
Specific Gravity 1.12
Synonym ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
Solubility Information Solubility in water: insoluble
Formula Weight 192.21
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethyl 3-phenylglycidate, 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'