Ethyl 3-phenylglycidate, 90%, mixture of cis and trans, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 332650010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 121-39-1 Weight: 1.82500kg


Packaging Glass bottle
Quantity 1L

Product Code. 10647884

Quantity Price
1 £ 238.2 / 1 L
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl 3-phenylglycidate
Name Note 90%
CAS 121-39-1
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C11H12O3
Formula Weight 192.21
MDL Number MFCD00005123

Specification

Assay ≥88%
Formula Weight 192.21
Physical Form Liquid
Quantity 1L
Color Clear colorless to yellow
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 204-467-3
Infrared Spectrum Authentic
CAS 121-39-1
RTECS Number MB4970000
Assay Percent Range 90%
Refractive Index 1.5165 to 1.5205 (20°C, 589nm)
MDL Number MFCD00005123
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Molecular Formula C11H12O3
Name Note 90%
Density 1.1200g/mL
Health Hazard 1 Warning
TSCA TSCA
Chemical Name or Material Ethyl 3-phenylglycidate
Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.