missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%

Brand:  Alfa Aesar™ H64533.03

Code : NEW

Additional Details : CAS Number : 26510-52-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491789

  • £80.10 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 3-oxo-3-(2-pyridyl)propionate
193.2
145706
26510-52-1
MFCD00094022
3-Oxo-3-(2-pyridyl)propionic acid ethyl ester; Ethyl picolinoylacetate

Specifications

Ethyl 3-oxo-3-(2-pyridyl)propionate
100° to 104°C (0.3mmHg)
26510-52-1
1.5160
96%
MFCD00094022
3-Oxo-3-(2-pyridyl)propionic acid ethyl ester; Ethyl picolinoylacetate
Light Sensitive
1g
1.164g/mL
Liquid
193.2
145706
Videos
SDS