missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-oxo-3-(2-pyridyl)propionate, 96%, Thermo Scientific™

Catalog Number 15401799
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
26510-52-1
C10H11NO3
193.20
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
2736461
CCOC(=O)CC(=O)C1=CC=CC=N1
This item is not returnable. View return policy
26510-52-1
C10H11NO3
193.20
MFCD00094022
FQHXWZMJALFSJJ-UHFFFAOYSA-N
ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
2736461
CCOC(=O)CC(=O)C1=CC=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.164 g/mL
Boiling Point 100°C to 104°C (0.3 mmHg)
Flash Point >200°C (392°F)
Refractive Index 1.516
Quantity 5 g
Beilstein 145706
Sensitivity Light sensitive
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
Formula Weight 193.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl 3-oxo-3-(2-pyridyl)propionate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'