missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%, Thermo Scientific Chemicals

Catalog Number p-128486
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
924-99-2
C7H13NO2
143.19
MFCD00144269
MVUMJYQUKKUOHO-AATRIKPKSA-N
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
5369162
ethyl (E)-3-(dimethylamino)prop-2-enoate
CCOC(=O)C=CN(C)C
This item is not returnable. View return policy
924-99-2
C7H13NO2
143.19
MFCD00144269
MVUMJYQUKKUOHO-AATRIKPKSA-N
ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
5369162
ethyl (E)-3-(dimethylamino)prop-2-enoate
CCOC(=O)C=CN(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17.0°C to 18.0°C
Color Orange to Yellow
Density 1.0000g/mL
Boiling Point 118°C to 121°C (10.0 HPA)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH=CHCOOC2H5
Refractive Index 1.5105 to 1.5125
Quantity 25 g
Specific Gravity 1
Solubility Information Solubility in water: 90g/L (25 c)
Formula Weight 143.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl 3-(N, N-dimethylamino)acrylate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-(N,N-dimethylamino)acrylate
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'