missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 3-methylisoxazole-5-carboxylate, 95%

Brand:  Alfa Aesar™ H63741.03

Code : NEW

Additional Details : CAS Number : 63366-79-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15440589

  • £85.75 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 3-methylisoxazole-5-carboxylate
155.15
3-Methylisoxazole-5-carboxylic acid ethyl ester
63366-79-0
MFCD11656615

Specifications

Ethyl 3-methylisoxazole-5-carboxylate
63366-79-0
95%
MFCD11656615
1g
Solid
155.15
3-Methylisoxazole-5-carboxylic acid ethyl ester
Videos
SDS