missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-fluorobenzoyl)acetate, 98%, Thermo Scientific Chemicals

Catalog Number 15497838
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
33166-77-7
C11H11FO3
210.204
MFCD03424809
MLABEWHVTXMKHP-UHFFFAOYSA-N
2063904
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC(=CC=C1)F
This item is not returnable. View return policy
33166-77-7
C11H11FO3
210.204
MFCD03424809
MLABEWHVTXMKHP-UHFFFAOYSA-N
2063904
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC(=CC=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Orange
Density 1.166 g/mL
Boiling Point 262°C to 263°C
Refractive Index 1.517
Quantity 250 mg
Synonym ethyl 3-3-fluorophenyl-3-oxopropanoate, ethyl 3-fluorobenzoyl acetate, 3-3-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-3-fluoro-phenyl-3-oxopropanoate, benzenepropanoic acid, 3-fluoro-beta-oxo-, ethyl ester, ethyl3-3-fluorophenyl-3-oxopropanoate, acmc-1cl0f, ethyl-m-fluorobenzoylacetate, ethyl 3'-fluorobenzoylacetate, ksc570a6p
Formula Weight 210.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl (3-fluorobenzoyl)acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'