missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxypropionate, 99+%, pure, stabilized, Thermo Scientific Chemicals

Catalog Number p-128557
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
763-69-9
C7H14O3
146.19
MFCD00051356
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
12989
ethyl 3-ethoxypropanoate
CCOCCC(=O)OCC
This item is not returnable. View return policy
763-69-9
C7H14O3
146.19
MFCD00051356
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
12989
ethyl 3-ethoxypropanoate
CCOCCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -75°C
Density 0.9490g/mL
Boiling Point 170°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC(O)CH2CH2OC2H5
Refractive Index 1.4060 to 1.408
Quantity 1 L
Specific Gravity 0.949
Solubility Information Solubility in water: 1.6% (20 c)
Stabilizer 50 to 100ppm BHT
Viscosity 1.3 mPa.s (20°C)
Formula Weight 146.19
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethyl 3-ethoxypropionate, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 3-ethoxypropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'