missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxyacrylate, 98%, Thermo Scientific Chemicals

Catalog Number 15455428
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1001-26-9
C7H12O3
144.17
MFCD00009863
ITQFPVUDTFABDH-AATRIKPKSA-N
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate
5366269
ethyl (E)-3-ethoxyprop-2-enoate
CCOC=CC(=O)OCC
This item is not returnable. View return policy
1001-26-9
C7H12O3
144.17
MFCD00009863
ITQFPVUDTFABDH-AATRIKPKSA-N
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate
5366269
ethyl (E)-3-ethoxyprop-2-enoate
CCOC=CC(=O)OCC

Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.998 g/mL
Boiling Point 195°C to 196°C
Flash Point 76°C (169°F)
Linear Formula CH3CH2OCH=CHCOOCH2CH3
Refractive Index 1.4465
Quantity 10 g
UN Number UN3334
Beilstein 1721612
Solubility Information Slightly soluble in water.
Formula Weight 144.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-ethoxyacrylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'