missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chloro-2,2-dimethylpropanoate, ≥97%, Thermo Scientific™

Catalog Number p-3110952
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
106315-37-1
C7H13ClO2
164.629
MFCD00220052
NEVHNBRNUWSFSB-UHFFFAOYSA-N
287572
ethyl 3-chloro-2,2-dimethylpropanoate
CCOC(=O)C(C)(C)CCl
This item is not returnable. View return policy
106315-37-1
C7H13ClO2
164.629
MFCD00220052
NEVHNBRNUWSFSB-UHFFFAOYSA-N
287572
ethyl 3-chloro-2,2-dimethylpropanoate
CCOC(=O)C(C)(C)CCl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-chloro-2,2-dimethyl-propionic acid ethyl ester, propanoic acid,3-chloro-2,2-dimethyl-, ethyl ester, acmc-1c8oo, ethyl 3-chloro-2,2-dimethyl-propanoate, propanoic acid, 3-chloro-2,2-dimethyl-, ethyl ester
Formula Weight 164.63
Percent Purity 97%
Chemical Name or Material Ethyl 3-chloro-2, 2-dimethylpropanoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'