missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-bromobenzoate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4'-methyl-biphenyl-3-carboxylic acid ethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4′-methyl-biphenyl-3-carboxylic acid ethyl ester.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4′-methyl-biphenyl-3-carboxylic acid ethyl ester.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Density | 1.43 |
Boiling Point | 261°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.543 |
Quantity | 10 g |
Beilstein | 1867121 |
Synonym | ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate |
Formula Weight | 229.07 |
Percent Purity | ≥98% |
Chemical Name or Material | Ethyl 3-bromobenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'