Learn More
Alfa Aesar™ Ethyl 3-aminopyridine-4-carboxylate, 95%
Brand: Alfa Aesar™ H64188.06
Code : NEW
Additional Details : CAS Number : 14208-83-4 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Ethyl 3-aminopyridine-4-carboxylate | |
166.18 | |
383705 |
14208-83-4 | |
MFCD03788270 | |
3-Aminopyridine-4-carboxylic acid ethyl ester; Ethyl 3-aminoisonicotinate |
Specifications
Ethyl 3-aminopyridine-4-carboxylate | |
Solid | |
62° to 64°C (lit.) | |
166.18 | |
383705 |
14208-83-4 | |
95% | |
5g | |
MFCD03788270 | |
3-Aminopyridine-4-carboxylic acid ethyl ester; Ethyl 3-aminoisonicotinate |