missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020884
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
626-34-6
C6H11NO2
129.16
MFCD00008073,MFCD02730138
YPMPTULBFPFSEQ-UHFFFAOYSA-N
z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
643756
ethyl (Z)-3-aminobut-2-enoate
CCOC(=O)C=C(C)N
This item is not returnable. View return policy
626-34-6
C6H11NO2
129.16
MFCD00008073,MFCD02730138
YPMPTULBFPFSEQ-UHFFFAOYSA-N
z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
643756
ethyl (Z)-3-aminobut-2-enoate
CCOC(=O)C=C(C)N

Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

Solubility
Soluble in water (26 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acids, bases, oxidizing agents, reducing agents. Air & Moisture Sensitive & Hygroscopic.
TRUSTED_SUSTAINABILITY
Melting Point 29°C to 35°C
Boiling Point 210°C to 215°C
Flash Point 97°C (206°F)
Quantity 25 g
UN Number UN3263
Beilstein 471284
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-aminocrotonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'