ethyl 3,5-dichloro-4-hydroxybenzoate, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: SPB00157FA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 17302-82-8 Weight: 0.15000kg


Packaging Amber glass bottle
Quantity 100g

Product Code. 10275904

Quantity Price
1 £ 56.0 / 100g
Estimated Shipment date
from Supplier 31-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethyl 3, 5-dichloro-4-hydroxybenzoate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8Cl2O3
Formula Weight 235.07

Specification

Formula Weight 235.07
Quantity 100g
Assay 97%
Molecular Formula C9H8Cl2O3
Assay Percent Range 97%
Packaging Amber glass bottle
CAS 17302-82-8
Chemical Name or Material Ethyl 3, 5-dichloro-4-hydroxybenzoate
Documentation

Technical documents