missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-diethoxypropanoate, 98%, Thermo Scientific Chemicals

Catalog Number p-3757856
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
10601-80-6
C9H18O4
190.24
MFCD00009865
SIALOQYKFQEKOG-UHFFFAOYSA-N
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
66389
ethyl 3,3-diethoxypropanoate
CCOC(CC(=O)OCC)OCC
This item is not returnable. View return policy
10601-80-6
C9H18O4
190.24
MFCD00009865
SIALOQYKFQEKOG-UHFFFAOYSA-N
ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
66389
ethyl 3,3-diethoxypropanoate
CCOC(CC(=O)OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9700g/mL
Boiling Point 95°C (17.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5O)2CHCH2COOC2H5
Refractive Index 1.4100 to 1.412
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: 23g/L (20 c)
Formula Weight 190.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3, 3-diethoxypropanoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'