missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate, 95%

Brand:  Alfa Aesar™ H61453.03

Code : NEW

Additional Details : CAS Number : 121195-03-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457998

  • £46.22 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate
222.26
3-(2-Thienyl)-1H-pyrazole-5-carboxylic acid ethyl ester
121195-03-7
MFCD07772880

Specifications

Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate
121195-03-7
95%
MFCD07772880
1g
Solid
222.26
3-(2-Thienyl)-1H-pyrazole-5-carboxylic acid ethyl ester
Videos
SDS