missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 3-(2-bromophenyl)propionate, 98%

Brand:  Alfa Aesar™ H63505.03

Code : NEW

Additional Details : CAS Number : 135613-33-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15420239

  • £18.60 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 3-(2-bromophenyl)propionate
257.13
3-(2-Bromophenyl)propionic acid ethyl ester
135613-33-1
MFCD07772962

Specifications

Ethyl 3-(2-bromophenyl)propionate
Liquid
Pale yellow
257.13
3-(2-Bromophenyl)propionic acid ethyl ester
135613-33-1
98%
1g
MFCD07772962
Videos
SDS