missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-[2,6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate, 97%, Thermo Scientific™

Catalog Number p-128522
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
96568-04-6
C10H8Cl2FNO3
280.08
MFCD00799535
IEUHWNLWVMLHHC-UHFFFAOYSA-N
ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate
2733708
ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl
This item is not returnable. View return policy
96568-04-6
C10H8Cl2FNO3
280.08
MFCD00799535
IEUHWNLWVMLHHC-UHFFFAOYSA-N
ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate
2733708
ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 72°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 280.08
Percent Purity 97%
Chemical Name or Material Ethyl 3-[2, 6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate, 97%
compliance-icons
Product Identifier
  • Ethyl 3-¢2,6-dichloro-5-fluoro-(3-pyridiyl)!-3-oxopropanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'