missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals
32.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Density | 1.064 |
Boiling Point | 106°C (11 mmHg) |
Flash Point | 96°C (205°F) |
Linear Formula | (O=)C6H9CO2C2H5 |
Refractive Index | 1.4775 |
Quantity | 25 g |
Beilstein | 608356 |
Solubility Information | Soluble in water. |
Formula Weight | 170.21 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'