missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(N-phthalimidooxy)-2-methylpropionate, 98%, Thermo Scientific™

Catalog Number p-7053903
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
40674-21-3
C14H15NO5
277.276
UNZUMKCIIIFEJI-UHFFFAOYSA-N
ethyl 2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methylpropionate, ethyl 2-1,3-dioxoisoindolin-2-yloxy-2-methylpropanoate, ethyl 2-1,3-dioxoisoindol-2-yl oxy-2-methylpropanoate, 2-phthalimidyloxy isobutyric acid ethyl ester, propanoic acid,2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methyl-, ethyl ester
3016209
ethyl 2-(1,3-dioxoisoindol-2-yl)oxy-2-methylpropanoate
CCOC(=O)C(C)(C)ON1C(=O)C2=CC=CC=C2C1=O
This item is not returnable. View return policy
40674-21-3
C14H15NO5
277.276
UNZUMKCIIIFEJI-UHFFFAOYSA-N
ethyl 2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methylpropionate, ethyl 2-1,3-dioxoisoindolin-2-yloxy-2-methylpropanoate, ethyl 2-1,3-dioxoisoindol-2-yl oxy-2-methylpropanoate, 2-phthalimidyloxy isobutyric acid ethyl ester, propanoic acid,2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl oxy-2-methyl-, ethyl ester
3016209
ethyl 2-(1,3-dioxoisoindol-2-yl)oxy-2-methylpropanoate
CCOC(=O)C(C)(C)ON1C(=O)C2=CC=CC=C2C1=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 277.27
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-(N-phthalimidooxy)-2-methylpropionate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'