missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate, tech. 90%

Brand:  Alfa Aesar™ H63971.03

Code : NEW

Additional Details : CAS Number : 2549-19-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498606

  • £136.74 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate
Technical Grade
MFCD00139509
2549-19-1
204.23
2-Methyl-imidazo[1,2-a]pyridine-3-carboxylic acid ethyl ester

Specifications

Ethyl 2-methylimidazo[1,2-a]pyridine-3-carboxylate
1g
Technical Grade
90%
MFCD00139509
54° to 57°C
2549-19-1
Solid
204.23
2-Methyl-imidazo[1,2-a]pyridine-3-carboxylic acid ethyl ester
Videos
SDS