missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylacetoacetate, 95%, Thermo Scientific Chemicals

Catalog Number p-130789
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
609-14-3
C7H12O3
144.17
MFCD00009164
FNENWZWNOPCZGK-UHFFFAOYSA-N
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
701
ethyl 2-methyl-3-oxobutanoate
CCOC(=O)C(C)C(=O)C
This item is not returnable. View return policy
609-14-3
C7H12O3
144.17
MFCD00009164
FNENWZWNOPCZGK-UHFFFAOYSA-N
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
701
ethyl 2-methyl-3-oxobutanoate
CCOC(=O)C(C)C(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -45°C
Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 187°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(CH3)CO2C2H5
Refractive Index 1.4170 to 1.419
Quantity 25 g
Beilstein 03, 679
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
Formula Weight 144.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-methylacetoacetate, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'